missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators

Product Code. 17232484 Shop All Thermo Scientific Products
Click to view available options
Detectable Analytes:
Vancomycin
This item is not returnable. View return policy

Product Code. 17232484

Brand: Thermo Scientific™ 0373597

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ TDM Calibrators, offered in Levels A to F.

Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.

Reduces Labor
  • Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
  • Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
Ready-to-use
  • Liquid stable and ready-to-use
  • Applicable to general chemistry analyzers
  • Validated parameters assure lab-to-lab reproducibility
  • Superb reagent stability
Optimal Sensitivity
  • Excellent correlation to reference methods
  • Low cross-reactivity with metabolites and commonly co-administered drugs
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Vancomycin
Sample Type Plasma, Serum
Type Calibrator
Quantity Levels A-F, 1 x 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.