Learn More
Hellma™ QS Quartz Suprasil High Precision Macro Absorption Cells With PTFE Stopper
Brand:  Hellma™ 110-1-40
 View more versions of this product
Product Code. 10475031
  • 284.96 € / Each
Explore more special offers
Specifications
Macro Absorption Cell
Quartz Suprasil
9.5mm
200 to 2500 nm
12.5 mm
350 μL
PTFE
1 mm
52 mm
Photometric and Colorimetric Measurements