Learn More
QST and Fragment Chemistry Installation Kit
GeneScan™ 500 ROX™ Size Standard is designed for sizing DNA fragments in the 35-500 bp range, provides 16 single-stranded ROX™ dye labeled fragments.
Brand: ABI Applied Biosystems 4413691
This item is not returnable.
View return policy
Description
GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems.Order Info
1 Kit
Specifications
GeneScan™ 500 ROX™ dye Size Standard, 800 rxns, ROX™ Label, Use With 310 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer | |
QST and Fragment Chemistry Installation Kit | |
QST and Fragment Chemistry Installation Kit |