missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ QSY™ 7 Carboxylic Acid, Succinimidyl Ester

QSY™ 7 carboxylic acid, succinimidyl ester

Brand:  Molecular Probes™ Q10193

374.68 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 11573657

  • 493.00 € / 5mg
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

QSY-7 succinimidyl ester is a nonfluorescent acceptor dye for preparation of peptide and oligonucleotide FRET probes via aliphatic amine modification.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mg
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
QSY™ 7
QSY™
Succinimidyl Ester
Amine
Active Ester, Succinimidyl Ester
Quencher Dyes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.