Learn More
BINDER™ Qualification Documents IQ/OQ
Qualification documents - IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, humidity, and light values
Brand: BINDER™ 7007-0001
This item is not returnable.
View return policy
Description
- Parameters depending on unit
- Available as hardcopy inside folder or PDF document
Specifications
BINDER Chambers Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V | |
On request | |
Supporting documents for validation performed by customers |
Qualification documents - Hard copy inside folder, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, CO2, O2, pressure, depending on unit | |
IQ/OQ Documentation (Hardcopy) | |
IQ/OQ checklists include calibration guide and comprehensive unit documentation |
Product Content Correction
Product Title
BINDER™ Qualification Documents IQ/OQ > IQ/OQ; version: hardcopy inside folder; parameters: temperature, CO2, O2, pressure, depending on unit
Spot an opportunity for improvement?Share a Content Correction