Learn More
BINDER™ Qualification Documents IQ/OQ
Qualification documents - Digital in PDF format, IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists including calibration guide and comprehensive unit documentation.
Brand:  BINDER™ 7057-0001
 View more versions of this product
Product Code. 16484009
  • 1120.00 € / Each
Explore more special offers
Description
Description
  • Parameters depending on unit
  • Available as hardcopy inside folder or PDF document
Specifications
Specifications
BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V)
IQ/OQ Documentation (PDF)
IQ/OQ/PQ with completion of documents
Qualification documents - Digital in PDF format, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, CO2, O2, pressure, depending on unit
BINDER Chambers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Qualification Documents IQ/OQ > IQ/OQ; For use with: BINDER Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V