Learn More
Socorex™ Qualitix™ Sterilization Pouch
Product Code. p-7084336
Click to view available options
Dimensions (L x W):
83 x 160 mm
90 x 260 mm
135 x 260 mm
190 x 360 mm
300 x 395 mm
Unit Size:
Pack of 200
Product Code. 15710679
Brand: Socorex™ 3210.0816
Only null left
Add to basket
Edge
Add to basket
Fits any small to mid-size sterilization device and provides easy and effective wrapping of items. Made of robust medical grade paper, they tightly close with self-sealing strip to maintain sterility. Content remains visible through PET/CPP transparent laminated film.
  • Robust durable material
  • Quick content identification
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
  • Conformity to ISO 17665-06, ISO 11135, CE
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO 17665-06, ISO 11135, CE
Closure Type Self-adhesive
Material Paper
Dimensions (L x W) 83 x 160 mm