Learn More
Sampling Systems™ Quality Control PharmaLabels
Medium strength adhesion paper labels with a special FDA acceptable adhesive. Sampling Systems™ Quality Control PharmaLabels are ideal for applications where the adhesive might come into contact with the product.
Brand:  Sampling Systems™ 8220P-04
 View more versions of this product
Product Code. 15829501
  • 66.45 € / Pack of 500
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Medium strength adhesive
  • Large writing area
  • Each roll individually packed
  • FDA acceptable adhesive (CFR 175.105)
  • Ideal for applications where the adhesive might come into contact with the product
Specifications
Specifications
CFR 175.105
Blue
PharmaLabel
500/Pk.
95 x 95 mm
Food Grade Adhesive
Paper
Quality Control
-40°C to +70°C