missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ OliGreen™ ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent

Quant-iT™ OliGreen ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent contain an ultrasensitive, green-fluorescent nucleic acid stain for quantitating oligonucleotides and single-stranded DNA (ssDNA) in solution.

920.00 € - 1006.00 €

Specifications

Detection Method Fluorescence
Excitation/Emission 500/525
For Use With (Equipment) Microplate Reader
No. of Reactions 200 assays (2 mL assay volume)
Quantitation Range 200 pg to 2 μg
View More Specs

Products 2
Product Code Brand Product Type Quantitation Range Price Quantity & Availability  
Product Code Brand Product Type Quantitation Range Price Quantity & Availability  
10492152
View accessory products View Documents
Invitrogen™
O11492
Quant-iT OliGreen ssDNA Kit 200 pg to 2 μg
1006.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10729443
View Documents
Invitrogen™
O7582
Quant-iT OliGreen ssDNA Reagent 200 pg to 2 μg
920.00 €
1mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Quantify ssDNA with ease using the Quant-iT OliGreen ssDNA Assay kit and reagent. This assay kit and reagent enable sensitive quantitation of single-stranded DNA without the need for a highly concentrated sample. As little as 100 pg/mL can be quantitated using the assay kit or Quant-iT OliGreen ssDNA Reagent.

Quantitate ssDNA and other oligonucleotides accurately and without the need for a highly concentrated sample with Quant-iT OliGreen ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent. Quant-iT OliGreen ssDNA Assay Kits contain an ultrasensitive, green-fluorescent nucleic acid stain for quantitating oligonucleotides and single-stranded DNA (ssDNA) in solution. The Quant-iT™ OliGreen™ ssDNA Assay Reagent enables researchers to quantify as little as 1 ng/mL oligonucleotide or ssDNA (200 pg in a 200 μL assay volume) with a fluorescence microplate reader using fluorescein excitation and emission wavelengths. This sensitivity exceeds that achieved with absorbance methods by 10,000-fold.

In addition to the dye, the OliGreen ssDNA Assay Kit includes buffer and an oligonucleotide standard. The OliGreen reagent is also available as the stand-alone Quant-iT OliGreen ssDNA Reagent.

Specifications

Specifications

Fluorescence
Microplate Reader
200 pg to 2 μg
Room Temperature
500/525
200 assays (2 mL assay volume)
OLIGREEN, Quant-iT
Videos
SDS
Documents

Documents

Product Certifications