missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuantaRed™ Enhanced Chemifluorescent HRP Substrate Kit

Enable sensitivity comparable to enhanced chemiluminescence (ECL) using the most sensitive fluorescent ELISA substrate for peroxidase (HRP).

Brand:  Thermo Scientific™ 15159


Product Code. 10053933

  • 498.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ QuantaRed Enhanced Chemifluorescent HRP Substrate is the most sensitive fluorescent ELISA substrate, providing sensitivity comparable to enhanced chemiluminescence.

QuantaRed Enhanced Chemifluorescent HRP Substrate enables high-yield and sensitive fluorescent and colorimetric detection of peroxidase activity in the ELISA format. QuantaRed Substrate uses a proprietary technology to increase fluorescent yield and sensitivity of the well-documented ADHP (10-acetyl-3,7-dihydroxyphenoxazine) chemifluorescence reaction. ADHP is a non-fluorescent compound that reacts with horseradish peroxidase (HRP) to produce resorufin, a soluble, highly fluorescent reaction product (excitation/emission approx. 570/585nm). The long wavelength emission of the resorufin minimizes interference from the low wavelength (blue and green) autofluorescence, making it an ideal substrate for quantitating target molecules in biological samples. In addition, the colored reactopm product of QuantaRed Substrate can be measured spectrophotmetrically at 570nm.

Highlights:

  • Sensitive – low picogram to femtogram detection

  • Red-shifted – less interference from autofluorescence generated by biological samples

  • Convenient – compatible with existing filter sets, no need to purchase special filter sets

  • Complete – easy to use; no buffers to prepare or reagent powders to dissolve

  • Flexible – effective stop solution result in stable signal for at least 4 hours

  • Versatile – produces strong signal for easy fluorometric or colorimetric measurement

Note:

Equilibrate all reagents to room temperature before use. Temperature can affect the reaction kinetics.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ELISA Substrate
Fluorescence
Chemical Substrate
Sufficient For: 960 wells (10 x 96-well plates) at 100 μL working solution per well
• QuantaRed ADHP Concentrate, 1 mL
• QuantaRed Enhancer Solution, 50 mL
• QuantaRed Stable Peroxide Solution, 50 mL
• QuantaRed Stop Solution, 10 mL

Store at 4°C.
110 ml
QuantaRed™
HRP (Horseradish Peroxidase)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • QuantaRed Enhanced Chemifluorescent HRP Substrate Kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Aliphatic carboxylicacid, trometamol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.