Learn More
Macherey-Nagel™ Quantofix™ Peroxide 100 Test Strip Kits
Allows for the quick and easy determination of peroxide levels in solutions. Macherey Nagel™ Quantofix™ Peroxide 100 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Brand:  Macherey-Nagel™ 91312
Product Code. 10159960
  • 78.90 € / Pack of 100
Edge
Explore more special offers
Description
Description
Allows for the quick and easy determination of peroxide levels in solutions. Macherey Nagel™ Quantofix™ Peroxide 100 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
  • Ready to use
  • Rapid: provides reliable results within 15 seconds
  • CE marked and suitable to be used in the healthcare environment
  • Can be used in chemical laboratories to check organic solvents
  • Test for residual peroxide in cleaning procedures with high potency disinfectants
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store at room temperature
Specifications
Specifications
White to Blue
Determination of Peroxide 100
100 Pack
15 sec.
Strip
0, 1, 3, 10, 30, 100mg/L H2O2
Room Temperature
Semi-qualitative Test Strip
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Quantofix™ Peroxide 100 Test Strip Kits > Color Indicator: White to Blue