Learn More
Applied Biosystems™ QuantStudio™ 6 Pro Real-Time PCR System, AB Assurance extended warranty, SmartStart, 384-well
The Applied Biosystems™ QuantStudio™ 6 Pro Real-Time PCR System Training and Extended Warranty Package adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint.
Brand:  Applied Biosystems™ A45582
Product Code. 17142106
  • 82509.00 € / Each
Explore more special offers
Description
Description
  • Easy and convenient hands-free operation with facial authentication and voice commands
  • Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
  • Space savings with small footprint and true standalone operation; set-up, start, monitor, and analyze directly on the touchscreen
  • Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • SmartStart Orientation gets users up and running quickly, connects them to their local network, and assists with choosing appropriate reagents/consumables
  • 1 year AB Assurance extended warranty including comprehensive service coverage and one planned maintenance visit
  • Service plans maximize system uptime with fast repair turnaround time from manufacturer-trained and certified field service engineers (FSEs)
  • During annual maintenance, the FSE will update software if required, clean, lubricate, and align relevant parts and optical systems, functional tests as well as document tasks, results, and findings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Interchangeable
Energy Efficient, Fewer Resources used and Less Waste
1 system
QuantStudio™
Room Temperature