missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL
The Applied Biosystems QuantStudio 7 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Package includes the QuantStudio 7 Pro Real-Time PCR System, SmartSmart Orientation, and a 1 or 3 year extended warranty.
Brand: Applied Biosystems™ A44557
This item is not returnable.
View return policy
Description
- Adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint
- The QuantStudio 7 Pro system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation
- Designed for ultimate ease-of-use, the system offers high quality, high throughput, and excellent reliability
- Easy and convenient hands-free operation with facial authentication and voice commands
- Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
- Space savings with small footprint and true standalone operation; setup, start, monitor, and analyze directly on the touchscreen
- Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- An optimal user experience for researchers who want to work smarter in the lab
- Package includes the QuantStudio 7 Pro Real-Time PCR System, SmartSmart Orientation, and a three year extended warranty
- Extended warranty includes comprehensive service coverage and one planned maintenance visit

Specifications
Interchangeable | |
Real Time Thermal Cyclers | |
Energy Efficient, Fewer Resources used and Less Waste | |
System + SmartStart + 3 yr ext warranty | |
1 Year Manufacturer Warranty and 1 Year of Field Service with PM |
QuantStudio 7 Pro Real Time PCR System, 96-well, 0.2 mL block with 3 year extended Warranty and SmartStart orientation | |
12 in. Color Touchscreen | |
Room Temperature | |
1 Day at Customer Site |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL > 1 day at customer site
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction