Learn More
Applied Biosystems™ QuantStudio™ Absolute Q Package, including Digital PCR System, desktop PC, Smart Start training and 1 Year extended warranty
The Applied Biosystems™ QuantStudio™ Absolute Q™ Digital PCR System combines the power of digital PCR (dPCR) absolute quantification with the simplicity of a qPCR workflow. With a single hands-on step that takes five minutes to complete, dPCR is now easier than ever.
Brand:  Applied Biosystems™ A53267
Product Code. 17417823
  • 98660.00 € / Each
Explore more special offers
Description
Description
The QuantStudio Absolute Q system provides a high quality, consistent, and easy-to-use solution for researchers who want to join the quantification revolution. Combining the power of digital PCR (dPCR) absolute quantification with the simplicity of a qPCR workflow, all the necessary steps for dPCR (compartmentalization, thermal cycling, and data acquisition) are integrated into a single instrument.
Features of the QuantStudio Absolute Q system include:
  • Easy and convenient qPCR-like workflow with only five minutes of hands-on time.
  • Reduced errors and manual inputs with all functions integrated into a single instrument.
  • Accomplish more, faster: Increased productivity with results in as little as 90 minutes.
  • QuantStudio Absolute Q Digital PCR software enables intuitive setup, monitoring, and analysis.
  • Security, auditing, and e-signature features for 21 CFR part 11-compliance support.
  • SmartStart Orientation is performed by a Field Application Scientist (FAS) remotely or in the lab.
  • Helps users quickly get up and running, connected to the local network/cloud, and choose appropriate reagents/consumable.
  • Reduced downtime with Smart Remote Support; show issues to Technical Support for speedy resolution.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Non-interchangeable
QuantStudio™ Absolute Q™ MAP16 Plate
Digital PCR System
Room Temperature
Digital PCR System, Desktop PC, Smart Start Training and 1 Year Extended Warranty
QuantStudio™
Digital PCR
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ QuantStudio™ Absolute Q Package, including Digital PCR System, desktop PC, Smart Start training and 1 Year extended warranty >