Learn More
Thermo Scientific Chemicals Quartz Discs
Manufactured by fusing naturally occurring crystalline silica
Brand:  Thermo Scientific Chemicals 042298.KS
64.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15436455
  • 65.22 € / Each
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mineral, Quartz Disc
Form: Ground and polished
50.8 mm
1.59 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific Chemicals Quartz Discs > 1each