missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Quartz Halogen Lamp

For use with BA210/310 Microscopes. Motic™ Quartz Halogen Lamp requires 6V/30W.

Brand:  Motic™ 1101002400452

Product Code. 15159404

  • 31.00 € / 1 unit
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • For use with BA210/310 Microscopes
  • Quartz Halogen
  • 6V
  • 30W

Order Info

dimProductType

Specifications

Specifications

Quartz Lamp
1 Ea.
30 w
BA210/310 Microscopes
6 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Motic™ Quartz Halogen Lamp > Product Type: Quartz Lamp

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.