Learn More
Thermo Scientific Chemicals Quartz microscope slide, fused, 50.8x25.4x1.0mm (2.0x1.0x0.0394in)
Fused quartz slides manufactured by fusing naturally occurring crystalline silica
Brand: Thermo Scientific Chemicals 042296.KG
58.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- High chemical purity, melting point and corrosion resistance
- Extremely hard with low coefficient of thermal expansion
- Features electrical insulation qualities and optical transmission from ultra-violet to infra-red
Specifications
50.8 mm | |
1 mm | |
1 Piece |
25.4 mm | |
Made from fused quartz | |
50.8 x 25.4 mm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.