Learn More
Hellma™ Quartz Suprasil Match Pair Dish
Brand:  Hellma™ 100-10-K-40
Product Code. 13429418
  • 320.00 € / Pack of 2
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Match Pair Dish
Quartz Suprasil
2Pack
200 to 2500 nm