Learn More
Hellma™ Quartz Suprasil Micro Cells
Brand:  Hellma™ 110-5-40
 View more versions of this product
Product Code. 10209951
  • 254.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Micro Cell
Quartz Suprasil
9.5mm
5 mm
46 mm
1.75 mL
PTFE
1Each
200 to 2500 nm
12.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products