Learn More
Hellma™ Quartz Suprasil Ultramicro Cell
Brand:  Hellma™ 1052101015-40
 View more versions of this product
Product Code. 12594557
  • 979.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Ultra-micro Cell
Quartz Suprasil
12.5 mm
5 μL
45 mm