missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ 4 System Verification Assay Kit

Qubit™ 4 System Verification Assay Kit is designed specifically for use with the Qubit™ 4 Fluorometer

Brand:  Invitrogen™ Q33237

66.50 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 16213001

  • 87.50 € / 50 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Fast, easy-to-use, reagent-based assay that tests the performance of the Qubit™ 4 Fluorometer
  • Provides sufficient reagents to perform up to 50 assays
  • Qubit™ 4 System Verification Assay Kit can only be used with the Qubit™ 4 Fluorometer.
  • Kit includes:
    • Blank reagent solution
    • Green fluorescent reagent - Designed to test the blue excitation (470 nm LED; 430 to 495 nm excitation filter) and green fluorescence detection (510 to 580 nm emission filter) on the fluorometer
    • Far-red fluorescent reagent - Designed to test the red excitation (635 nm LED; 600 to 645 nm excitation filter) and far-red fluorescence detection (665 to 720 nm emission filter)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Qubit™
Room Temperature
50 assays
Store at room temperature.
System Verification Assay Kit
Qubit 4 Fluorometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3086979 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberQ33237
Lot Number2896531 Certificate TypeCertificate of Analysis Date30/04/2024 Catalog NumberQ33237
Lot Number2714507 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog NumberQ33237
Lot Number2570376 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog NumberQ33237
Lot Number2510661 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog NumberQ33237
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Qubit™ 4 System Verification Assay Kit > 4 System Verification Assay Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.