Learn More
Invitrogen™ Qubit and Quant-it Assay Kit Standards
Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards.
Brand: Invitrogen™ Q33235
59.85 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards. The Qubit standards can be used in conjunction with different Qubit or Quant-iT nucleic acid quantitation assay kits and offer stand-alone versions of the kit standard for performing additional assays.
Qubit 1X dsDNA HS Assay Lambda Standard
Qubit 1X dsDNA HS Assay Lambda Standard can be used with either Qubit 1X dsDNA HS Assay Kits, Qubit dsDNA HS Assay Kits, or their corresponding Quant-iT Assay Kits. The standards are supplied at the same concentration as the highest concentration dsDNA standard (10 ng/μL) in the Quant-iT Assay Kits and can be diluted into the standard series.
Qubit 1X dsDNA BR Standards
Qubit 1X dsDNA BR (Broad Range) Standards can be used with either Qubit 1X dsDNA BR Assay Kits, Qubit dsDNA BR Assay Kits, or their corresponding Quant-iT Assay Kits. The Qubit 1X dsDNA BR Standards includes standards at two concentrations, 0 and 100 ng/μL. The high standard corresponds to the same concentration as the highest concentration dsDNA standard in the Quant-iT Assay Kits and can be diluted into the standard series.
Qubit RNA IQ Assay Standards
Qubit RNA IQ Assay RNA standards are designed for use with the Qubit RNA IQ Assay Kit. Sufficient amounts of the small and large and/or structured RNA standard are supplied for generating an IQ standard curve for up to 500 samples. The RNA IQ standards are designed to measure RNA degradation and are not interchangeable with the standards from other RNA quantitation assays, including the Qubit RNA HS, BR, or XR assay kits.
Note: Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex fluorometer.

Specifications
Qubit™ RNA IQ Assay – Small RNA Standard, 2 x 1.5 mL at 100 ng/μL in 1 mM citrate buffer Qubit™ RNA IQ Assay – Large RNA Standard, 10 x 300 μL at 100 ng/μL in 1 mM citrate buffer Store at ≤-70°C |
|
Qubit 4 Fluorometer, Qubit Flex Fluorometer | |
Qubit RNA IQ Standards | |
Dry Ice |
RNA Quantification | |
Qubit | |
1 set |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.