Learn More
Invitrogen™ Qubit™ Assay Reservoirs
Reservoirs that facilitate the repeated multi-channel of Qubit™ Reagents.
Brand:  Invitrogen™ Q33253
Product Code. 16448283
  • 216.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Facilitates the repeated multi-channel pipetting of Qubit™ reagents into Qubit™ Flex Assay Tube Strips
  • For use with Qubit™ Flex Fluorometer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Qubit Flex Fluorometer
DNA, RNA or Protein Quantitation