missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ dsDNA Quantification Assay Kits

Achieve accurate and precise quantification of double-stranded DNA with Qubit dsDNA HS and BR Assay Kits. These dsDNA quantification kits enable quick and selective detection of low and high abundance DNA samples, and can distinguish dsDNA from ssDNA, RNA, protein, and free nucleotides.

Brand:  Invitrogen™ Q32850

107.92 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10625443

  • 149.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: BR Assay Kit
This item is not returnable. View return policy

Description

Description

Achieve accurate and precise quantification of dsDNA with Qubit dsDNA BR (Broad Range) Assay Kits. These dsDNA quantification kits enable quick and selective detection of low and high abundance DNA samples, and can distinguish dsDNA from ssDNA, RNA, protein, and free nucleotides. Contaminants such as salts, solvents, or detergents are well-tolerated.

Qubit dsDNA HS and BR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA), RNA, protein, and free nucleotides. All kits provide concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a Qubit Fluorometer.

Qubit dsDNA HS Assay Kit

The Qubit dsDNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of sensitive DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.005 to 120 ng/μL, providing a detection range of 0.1-120 ng.

Qubit dsDNA BR Assay Kit

The Qubit dsDNA BR (Broad-Range) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 2,000 ng/μL, providing a detection range of 4-2,000 ng.

Notes

  • Qubit dsDNA HS and BR Assay kits can be used with any Qubit Fluorometer
  • Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

dsDNA Quantitation, Broad Range
510/527
100 Reactions
Qubit
Room Temperature
Fluorescence
Qubit Fluorometer
4 to 2000 ng
100 assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2989150 Certificate TypeCertificate of Analysis Date08/10/2024 Catalog NumberQ32850_LITL18 PROMO,Q32850
Lot Number2935438 Certificate TypeCertificate of Analysis Date17/07/2024 Catalog NumberQ32850_LITL18 PROMO,Q32850
Lot Number2892761 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog NumberQ32850_LITL18 PROMO,Q32850
Lot Number2856049 Certificate TypeCertificate of Analysis Date29/02/2024 Catalog NumberQ32850_LITL18 PROMO,Q32850
Lot Number2806854 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog NumberQ32850_LITL18 PROMO,Q32850
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Qubit™ dsDNA Quantification Assay Kits > Quantity: 1 Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.