missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ Endotoxin Detection Assay Kit

The Invitrogen Qubit Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on the Qubit Flex Fluorometer that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples.

Brand:  Invitrogen™ Q32891

357.20 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 17845247

  • 470.00 € / 80 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Invitrogen Qubit Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on the Qubit Flex Fluorometer that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples. Amebocyte lysates are widely used as a simple and sensitive assay for the detection of endotoxin lipopolysaccharide (LPS), the membrane component of gram-negative bacteria.

Features of the Qubit Endotoxin Detection Assay Kit include:
Highly sensitive with a broad range—detect as little as 0.01 EU/mL to 10.0 EU/mL
Specific—no interference from ß-glucans and suitable for wide range of samples, including protein, vaccine, plasmid, DNA, and RNA
Easy to use—when paired with the Qubit Flex Fluorometer, calculations are performed automatically, reducing the potential for error

Endotoxin levels in the samples are accurately determined using the included endotoxin standard of known concentration that is derived from E. coli strain O111: B4. Determining endotoxin levels is important to assess the efficiency of endotoxin removal methods and prevent endotoxic shock, inflammation, and/or sepsis in tissue culture cells and animals injected with endotoxin-contaminated proteins.

When endotoxin encounters the amebocyte lysate, a series of enzymatic reactions results in the activation of Factor C, Factor B, and pro-clotting enzyme. The activated enzyme catalyzes a cleavage event in the substrate to produce a strong fluorescent signal. After stopping the reaction, the resulting signal is measured on the Qubit Flex Fluorometer. The correlation between fluorescent signal is proportional to the endotoxin concentration in the sample.

Notes
• Designed for use with the Qubit Flex Fluorometer (Cat. No. Q33327) and not currently for use on Qubit 4 Fluorometers
• Optimized for use with Qubit Flex Pyrogen Free Assay Tube Strips (Cat. No. Q32893)
• Use the Qubit Flex System Verification Assay Kit (Cat. No. Q33254) prior to first assay run

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Endotoxin
Kit
Qubit Flex Fluorometer
80 reactions
Fluorescence
Protein, Antibody, Plasmid, DNA, RNA
80 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2994092 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberQ32891
Lot Number2936392 Certificate TypeCertificate of Analysis Date26/08/2024 Catalog NumberQ32891
Lot Number2907368 Certificate TypeCertificate of Analysis Date22/05/2024 Catalog NumberQ32891
Lot Number2861849 Certificate TypeCertificate of Analysis Date03/04/2024 Catalog NumberQ32891
Lot Number2785527 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog NumberQ32891
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Qubit™ Endotoxin Detection Assay Kit > 80 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.