Learn More
Invitrogen™ Qubit™ Flex Pyrogen-Free Tube Strips
Invitrogen Qubit Flex Pyrogen Free Tube Strips are clear 8 x 200-μL thin-walled tubes with attached flat caps.
Brand: Invitrogen™ Q32893
213.56 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Invitrogen Qubit Flex Pyrogen Free Tube Strips are clear 8 x 200-μL thin-walled tubes with attached flat caps designed for use with the Qubit Flex Fluorometer (Cat. No. Q33327) and the Qubit Endotoxin Detection Assay Kit (Cat. No. Q32891).
Notes
• For DNA, RNA, and protein quantification use Qubit Flex Assay Tube Strips (Cat. No. Q33252).
Specifications
Assay Tube Strip | |
120 strips | |
Qubit Flex Fluorometer |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.