Learn More
Invitrogen™ Qubit™ microRNA Assay Kits
Achieve accurate miRNA quantitation, even in the presence of rRNA, large mRNA, or contaminants, with the Qubit microRNA Assay Kit, which offers 100 and 500 assay formats.
Brand:  Invitrogen™ Q32881
 View more versions of this product
Product Code. 15401563
  • 459.00 € / 1 set
Explore more special offers
Description
Description
Achieve accurate and precise quantification of small RNA (∼20 nucleotides or base pairs) with Qubit miRNA (microRNA) Assay Kits. These miRNA quantification kits enable quick detection of all types of small RNA, including microRNA and siRNA, and both single stranded and double stranded RNA. The assay is highly selective for small RNA over rRNA or large mRNA, and can tolerate contaminants such as salts, solvents, or detergents.
The Qubit miRNA Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of microRNA samples. This assay is highly selective for miRNA over rRNA or large mRNA (>1000 bp). Depending on sample volume, the assay kit is designed to be accurate for initial miRNA sample concentrations of 0.025 to 150 ng/μL, providing a detection range of 0.5−150 ng.
The Qubit microRNA Assay Kit provides concentrated assay reagent, dilution buffer, and pre-diluted miRNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using the Qubit Fluorometer.
Notes
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fluorescence
microRNA Quantitation
Qubit
500 assays
498/518
Qubit Fluorometer
500 Reactions
microRNA Assay Kit
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3050048 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog NumberQ32881
Lot Number2872000 Certificate TypeCertificate of Analysis Date27/03/2024 Catalog NumberQ32881
Lot Number2566191 Certificate TypeCertificate of Analysis Date17/07/2023 Catalog NumberQ32881
Lot Number2507896 Certificate TypeCertificate of Analysis Date10/10/2022 Catalog NumberQ32881
Lot Number2486582 Certificate TypeCertificate of Analysis Date23/05/2022 Catalog NumberQ32881
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Qubit™ microRNA Assay Kits > 500 assays