missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuEChERS AOAC 2007.01 Method Clean-up Kits

Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Clean-up Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.

Brand:  Thermo Scientific™ S2-2-P-AOAC-KIT

 View more versions of this product

Product Code. 17329862

  • 204.92 € / Pack of 100
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ QuEChERS Clean-up Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

QuEChERS AOAC 2007.01 Method Clean-up Kits

These kits use dispersive solid phase extraction (dSPE) to purify supernatants obtained during extraction. Clean-up kits are available in prefilled 2 mL and 15 mL tube formats for the following matrices: general fruits and vegetables, pigmented fruits and vegetables, fruits and vegetables containing fats and waxes, and all other food types.

Chemicals used in the AOAC 2007.01 Method

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
  • NaOAc: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

QuEChERS AOAC Clean-up Kit; 50 mg PSA, 50 mg GCB, 150 mg MgSO4 prefilled in 2 mL Tubes; 100/Pk.
Salts in Prefilled Tubes
50 mg PSA, 50 mg GCB, 150 mg MgSO4
2 mL
QuEChERS Clean-up Kit
QuEChERS sample preparation for AOAC 2007.01 Method
Pigmented Fruits and Vegetables
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers