missing translation for 'onlineSavingsMsg'
Learn More
Quetiapine hemifumarate, Tocris Bioscience™ Product Code.: 30001650

Quetiapine hemifumarate, Tocris Bioscience™

Product Code. 30001650
50 mg
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy

Product Code. 30001650

Brand: Tocris Bioscience 4735/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-HT2/D2 antagonist; atypical antipsychotic

Atypical antipsychotic. Displays antagonist activity at serotonin (5-HT2) and dopamine (D2) receptors (IC50 values are 148 and 329 nM respectively). Lacks affinity at either D1 or muscarinic receptors.

Specifications

CAS 111974-72-2
Molecular Formula C29H33N3O10S
MDL Number MFCD03423782
Quantity 50 mg
Synonym quetiapine fumarate, 2-methoxyethanol; quetiapine
InChI Key JLWSQVHJLLYDPX-LVEZLNDCSA-N
SMILES OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C1=NC2=CC=CC=C2SC2=CC=CC=C12
IUPAC Name bis((2E)-but-2-enedioic acid); 2-[2-(4-{2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-10-yl}piperazin-1-yl)ethoxy]ethan-1-ol
Molecular Weight (g/mol) 615.65
PubChem CID 45358180
Formula Weight 441.54
Percent Purity >99%
Chemical Name or Material Quetiapine hemifumarate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Quetiapine hemifumarate, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.