Learn More
Thermo Scientific™ QuickBag™ Buffered Peptone Water (BPW) (ISO)
Shorten your workflow with easy-to-prepare 2.7L or 4.5L media bags: simply merge the segments to reconstitute in two minutes.
90.90 € - 91.85 €
Specifications
Content And Storage | 10°C to 30°C |
---|---|
Format | Convenience Format |
Product Type | Pre-enrichment Medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15755493
|
Thermo Scientific™
DQ1049A |
3 x 2.7 L |
90.90 €
Pack of 3 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
15765493
|
Thermo Scientific™
DQ1049B |
2 x 4.5 L |
91.85 €
Pack of 2 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
Description
Save time in preparing selective enrichment medium for the isolation of Salmonella spp. from food samples using Thermo Scientific™ QuickBag™ Buffered Peptone Water (ISO), which conforms to ISO 6579:2002. The irradiated medium is supplied in transparent plastic bags—simply pop the seal to mix the water and medium segments to reconstitute in two minutes. QuickBag Enrichment Media are available in 2.7L and 4.5L sizes to suit your workflow. Medium can be dispensed through the outlet tube as required.
Shorten your workflow with QuickBag UVM1 Broth. QuickBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.
Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.
Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which QuickBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.
Easy-to-use: fill and dispense with minimum training.
Energy saving: gamma irradiated, eliminating the need for costly thermal processing.
Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.
Reduces waste: unprepared QuickBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.
Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.
Space-saving: conveniently packed in flat boxes, QuickBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.
Specifications
10°C to 30°C | |
Convenience Format | |
Pre-enrichment Medium |
Safety and Handling
missing translation for 'shelfLife' : 2 years (rehydrated 10 Days)