Learn More
Pyrex™ Quickfit™ Arsenic Determincation Apparatus
Constructed of borosilicate glass. Pyrex™ Arsenic Determination Apparatus provides a method of analysis for arsenic and antimony determination.
Brand:  Pyrex™ 1AD/1
237.01 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 11991578
  • 298.00 € / Each
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Constructed of borosilicate glass. Pyrex™ Arsenic Determination Apparatus provides a method of analysis for arsenic and antimony determination.
  • Designed for the estimation of arsenic and antimony
  • Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 538nm or 5380Å.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Arsenic Determination Apparatus
Borosilicate Glass (3.3)
Formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 538nm or 5380Å.
1 Each