Learn More
Quickfit™ Borosilicate Glass Büchner Vacuum Filtration Funnels with Glass Sintered Disc
Useful where conventional filter papers would be destroyed by acids and alkalis
Brand:  Quickfit VB40/08M
 View more versions of this product
Product Code. 15598000
  • 495.00 € / Pack of 5
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from borosilicate glass for highest resistance to chemical attack
  • With Quickfit ground glass cone to allow direct filtration into Quickfit Büchner flasks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
80 mL
52 mm
24/29
40mm
4
Borosilicate Glass
ISO 3585, DIN 12217, ASTM E-438
For filtering solutions that can be filtered by filter papers which are not resistant to certain acids and bases
Quickfit™ ground Glass cone
Buchner Vacuum Filtration Funnel