Learn More
Quickfit™ Borosilicate Glass Cylindrical Culture Vessel
Manufactured from temperature resistant borosilicate glass. Quickfit™ Borosilicate Glass Cylindrical Culture Vessel is ideal for fermentation and culture work. Autoclavable with precision ground flat-flange joint. For use with Quickfit™ flat-flange lids and retaining clips.
Brand: Quickfit FV10L
2289.50 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Manufactured from temperature resistant borosilicate glass. QuickfitTM Borosilicate Glass Cylindrical Culture Vessel is ideal for fermentation and culture work. Autoclavable with precision ground flat-flange joint. For use with QuickfitTM flat-flange lids and retaining clips.
- Ideal for fermentation and culture work
- Manufactured from temperature resistant borosilicate glass
- Autoclavable
- With precision ground flat-flange joint
- For use with Quickfit™ flat-flange lids and retaining clips
Specifications
Flat | |
Borosilicate Glass | |
1 Each | |
227 mm |
10,000 mL | |
Quickfit™ flat flange lids and retaining clips, ideal for fermentation and culture work | |
375 mm |