missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Dreschel Bottle
Manufactured from borosilicate glass for corrosion-free performance. Quickfit™ Borosilicate Glass Dreschel Bottles provide an inexpensive method for washing or drying gases. The robust, durable construction guards against accidental breakage.
Brand: Quickfit MF29/3/250
Description
Manufactured from borosilicate glass for corrosion-free performance. Quickfit™ Borosilicate Glass Dreschel Bottles provide an inexpensive method for washing or drying gases. The robust, durable construction guards against accidental breakage.
- For use with Quickfit™ bottle heads
- Borosilicate glass for corrosion free performance
- Robust construction
Specifications
250 mL | |
Dreschel Gas Washing Bottle | |
Flat base: 102 mm | |
1 Pack |
Borosilicate Glass | |
24/29 | |
200 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction