missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Dreschel Bottle

Manufactured from borosilicate glass for corrosion-free performance. Quickfit™ Borosilicate Glass Dreschel Bottles provide an inexpensive method for washing or drying gases. The robust, durable construction guards against accidental breakage.

Brand:  Quickfit MF29/3/125

 View more versions of this product

Product Code. 11382055

  • 44.85 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Manufactured from borosilicate glass for corrosion-free performance. Quickfit™ Borosilicate Glass Dreschel Bottles provide an inexpensive method for washing or drying gases. The robust, durable construction guards against accidental breakage.

  • For use with Quickfit™ bottle heads
  • Borosilicate glass for corrosion free performance
  • Robust construction
TRUSTED_SUSTAINABILITY
Specifications

Specifications

125 mL
Dreschel Gas Washing Bottle
Flat base: 102 mm
1 Pack
Borosilicate Glass
24/29
163 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.