Learn More
Quickfit™ Borosilicate Glass Flange Lid
Constructed of Borosilicate Glass. Quickfit™ Borosilicate Glass Flange Lid includes precision ground flat-flange joints. Fits Quickfit™ wide neck reaction flasks and culture vessels. Each lid with one or more sockets to accept other Quickfit™ components.
Brand:  Quickfit MAF4/41
 View more versions of this product
Product Code. 10714537
  • 218.00 € / Each
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Constructed of Borosilicate Glass. Quickfit™ Borosilicate Glass Flange Lid is comprised of precision ground flat-flange joints. Fits Quickfit™ wide neck reaction flasks and culture vessels. Each lid includes one or more sockets to accept other Quickfit™ components.
  • With precision ground flat-flange joints
  • Fits Quickfit™ wide neck reaction flasks and culture vessels
  • Each lid with one or more sockets to accept other Quickfit™ components
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0°, 5°, 10°, Side Socket: 14/23, 15° Side Socket: 29/32, Vertical Center Socket: 29/32
Wide neck flasks
Flange lid
Borosilicate Glass
Sockets for use with other Quickfit™ components
100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Flange Lid > Flange Bore: 100mm; Vertical Center Socket: 29/32; Side sockets: 14/23 at 0deg., 14/23 at 5deg., 14/23 at 10deg., 29/32 at 15deg.