missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Inland Revenue Condenser

Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Inland Revenue Condenser is highly efficient. Vapour and distillate move down through the coil while coolant water flows outside the coil. Complete with detachable plastic hose connectors for the easy and safe attachment of flexible tubing.

Brand:  Quickfit C6/13/SC

275.31 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 12029702

  • 300.00 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Inland Revenue Pattern
  • Highly efficient
  • Vapour and distillate move down through the coil
  • Coolant water flows outside the coil
  • Complete with detachable plastic hose connectors for the easy and safe attachment of flexible tubing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Surface Area X 102m2: 2.5
24/29
Inland Revenue Condenser
205 mm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.