missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Iodine Flask
Complies with BS 2735
222.00 € - 897.00 €
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
11333714
|
Quickfit
FIB250/3 |
250 mL | 24/29 |
222.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
12959333
|
Quickfit
FIB500/3 |
500 mL | 24/29 |
243.00 €
Pack of 2 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||
15507850
|
Quickfit
FIB500/4 |
500 mL | 29/32 |
897.00 €
Pack of 10 |
Estimated Shipment: 18-03-2025
Log in to see stock. |
||||
15597840
|
Quickfit
FIB250/4 |
250 mL | 29/32 |
836.00 €
Pack of 10 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||
Description
Complies with BS 2735
- Complies with BS 2735
- For the determination of iodine
- Flask has ground socket and cup shaped top
- Stopper has extended handle which projects well above the liquid seal
Note: Stopper is not included; sold separately.
Specifications
BS 2735 | |
Top style: Cup shaped |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title