Learn More
Quickfit™ Borosilicate Glass Large Capacity Soxhlet Extractor
With flat flange joints to permit introduction of large samples
Brand:  Quickfit EX5/75
897.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 15507810
  • 984.00 € / Pack of 2
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conforms to BS 2071
  • Large capacity extractors
  • With flat flange joints to permit introduction of large samples
  • Used in conjuction with flat flange lid and retaining clip which should be ordered separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flat Flange Lid and Retaining Clip
600 mL
34/35
370 mm
Soxhlet Extractor
Pyrex Borosilicate Glass
BS 2071