Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
Brand:  Quickfit C1/23/SC
66.07 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11321855
  • 69.55 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
  • Complies with DIN 12576
  • Popular water-jacketed condenser suitable for distillation and reflux applications
  • With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN 12576
535 mm
Borosilicate Glass
Suitable for distillation and reflux applications
24/29
Liebig Condenser
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Liebig Condenser > Cone Size: 24/29; Length: 535mm; Effective Length: 395mm