Learn More
Quickfit™ Borosilicate Glass Reversible Condenser Coil
Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Reversible Condenser Coil is fitted on each end with a different size cone to reduce the need for adapters. Includes detachable plastic connectors for the attachment of flexible tubing.
Brand:  Quickfit CX3/35/SC
 View more versions of this product
Product Code. 15557780
  • 221.00 € / Pack of 2
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Highly versatile reflux condenser
  • Each end of the condenser is fitted with a different size cone thus reducing the need for adapters
  • With detachable plastic connectors for the attachment of flexible tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
335 mm (Overall) 160 mm (Effective)
24/29 and 34/35
Borosilicate Glass
Condenser Coil