Learn More
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
Brand:  Quickfit FF500/2S
 View more versions of this product
Product Code. 15547840
  • 662.00 € / Pack of 20
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
  • Complies with ISO 1773 and ISO 4797
  • With highest quality Quickfit™ ground glass socket
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • Flat base allows use with hotplates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 1773, ISO 4797
Borosilicate Glass
Short
Flat
500 mL
135 mm
19/26