Learn More
Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
93.40 € - 489.00 €
Product Code | Brand | For Use With (Equipment) | Description | Porosity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Description | Porosity | Price | Quantity & Availability | |||
10637615
|
Quickfit
MF48/13/250 |
For model MF29/3/250 Dreschel bottle | Stem length: 165 mm | 1 |
105.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||
11362065
|
Quickfit
MF48/23/500 |
For model MF29/3/500 Dreschel bottle | Stem length: 217 mm | 2 |
97.80 €
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||
11332065
|
Quickfit
MF48/13/125 |
For model MF29/3/125 Dreschel bottle | Stem Length: 132 mm | 1 |
93.70 €
Each |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||
11342065
|
Quickfit
MF48/13/500 |
For model MF29/3/500 Dreschel bottle | Stem Length: 217 mm | 1 |
93.50 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||
11322065
|
Quickfit
MF48/03/500 |
For model MF29/3/500 Dreschel bottle | Stem Length: 217 mm | 0 |
97.65 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||
11312065
|
Quickfit
MF48/03/250 |
For model MF29/3/250 Dreschel bottle | Stem Length: 165 mm | 0 |
93.40 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||
11302065
|
Quickfit
MF48/03/125 |
For model MF29/3/125 Dreschel bottle | Stem Length: 132 mm | 0 |
93.40 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||
15547940
|
Quickfit
MF48/23/250 |
For model MF29/23/250 Dreschel bottle | Stem Length: 165 mm | 2 |
488.00 €
Pack of 5 |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||
15557940
|
Quickfit
MF48/33/250 |
For model MF29/3/250 Dreschel bottle | Stem Length: 165 mm | 3 |
489.00 €
Pack of 5 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||
Description
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
- Complies with BS 2461
- Sintered dip tube disperses gas into bubbles for greater efficiency
- Choice of sinter porosities to achieve fine bubble size
Specifications
BS 2461 | |
Borosilicate Glass |