Learn More
Quickfit™ Borosilicate Glass Soxhlet Extractor
Conforms to BS 2071. Quickfit™ Borosilicate Glass Soxhlet Extractor removes soluble components or impurities from solids via repeated soaking/siphoning with solvent. A wide choice of extractors is available to accommodate most sizes of sample and thimbles. Accurately formed concentric siphon tube provides reliable performance.
Brand:  Quickfit EX5/83
149.15 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11381885
  • 164.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Conforms to BS 2071. Quickfit™ Borosilicate Glass Soxhlet Extractor removes soluble components or impurities from solids via repeated soaking/siphoning with solvent. A wide choice of extractors is available to accommodate most sizes of sample and thimbles. Accurately formed concentric siphon tube provides reliable performance.
  • Conforms to BS 2071
  • For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
  • Wide choice of extractors to accommodate most sizes of sample and thimbles
  • Accurately formed concentric siphon tube for reliable performance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Soxhlet Extractor
Pyrex Borosilicate Glass
Ideal for extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
50/42
200 mL
24/29
BS 2071
250 mm