Learn More
Quickfit™ Borosilicate Glass Still Head Adapter
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
50.80 € - 300.00 €
Specifications
For Use With (Application) Connecting flask and condenser
Type Still Head Adapter
Material Borosilicate Glass
View More Specs
Products 7
Product Code Brand Standard Taper Joint Price Quantity & Availability  
Product Code Brand Standard Taper Joint Price Quantity & Availability  
11972098
View Documents
Quickfit
SH4/1
Cone Size: 14/23 (Condenser), 14/23 (flask); Socket Size: 14/23
56.60 €
Each
12068483
View Documents
Quickfit
SH4/22
Cone Size: 19/26 (Condenser), 19/26 (flask); Socket Size: 14/23
50.80 €
Each
11332115
View Documents
Quickfit
SH4/2
Cone Size: 19/26 (Condenser), 19/26 (flask); Socket Size: 19/26
62.05 €
Each
12088483
View Documents
Quickfit
SH4/33
Cone Size: 24/29 (Condenser), 24/29 (flask); Socket Size: 14/23
67.25 €
Each
12078483
View Documents
Quickfit
SH4/23
Cone Size: 19/26 (Condenser), 24/29 (flask); Socket Size: 14/23
54.75 €
Each
12098483
View Documents
Quickfit
SH4/44
Cone Size: 29/32 (Condenser), 29/32 (flask); Socket Size: 14/23
74.70 €
Each
15527970
View Documents
Quickfit
SH4/55
Cone size: 34/35 (flask), 34/35 (condenser); Socket Size: 14/23 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
  • Used in distillation assemblies for connecting flasks to condenser
  • Upper socket designed to accept thermometer either via cone/screw thread adapter or thermometer pocket
Specifications
Specifications
Connecting flask and condenser
Still Head Adapter
Borosilicate Glass