Learn More
QuickFit™ Borosilicate Glass Test Tubes with Ground Socket
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
Brand: Quickfit MF24/2
This item is not returnable.
View return policy
Description
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
- Presicion-ground socket ensures exact fit for stopper
- Test tube sizes correspond with Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers
- Order tubes and stoppers separately
Specifications
Ground Socket | |
19 mL | |
Borosilicate Glass | |
5 Pack |
Stopper Size: 19/26 | |
100 mm | |
Quickfit Hollow Stopper, Fisher Scientific product code 12568855 | |
23 mm |