Learn More
Quickfit™ Two-Neck Round-Bottom Glass Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
Brand:  Quickfit FR250/3S/2A
 View more versions of this product
Product Code. 12070653
  • 71.10 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
  • With highest-quality Quickfit™ ground glass socket
  • Side neck useful for introducing gas inlet tubes and thermometers etc., into the flask
  • Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mL
130 mm
24/29 (Centre), 19/26 (side)
Borosilicate Glass
2
Round