Learn More
Quickfit™ Chromatography Columns with Rotaflo™ Stopcock and Ground Cone
Manufactured from borosilicate glass. Quickfit™ Chromatography Columns with Rotaflo™ Stopcock and Ground Cone includes glass sintered disc (porosity 0) to support column packing. GP Rotaflo™ stopcock with PTFE key provides fine control of eluent flow. Stem within lower cone allows mounting on a Büchner flask for vacuum flash chromatography.
Brand:  Quickfit CR12/10
1482.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 15527770
  • 1600.00 € / Pack of 5
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With glass sintered disc (porosity 0) to support column packing
  • GP Rotaflo™ stopcock with PTFE key for fine control of eluent flow
  • Stem within lower cone allows mounting on a Büchner flask for vacuum flash chromatography
  • Manufactured from borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chromatography Column
10 mm
Pyrex™ Borosilicate Glass
GP Rotaflo™ stopcock with PTFE key, Glass sintered disc
1 Pack
270 mm
19/26 Ground cone