Learn More
Quickfit™ Distillation Splash Head Adapter
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
Brand:  Quickfit SH7/12
93.39 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12078493
  • 101.00 € / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
A splash head for use in distillation assemblies in which the condenser is assembled in the vertical mode - Lower cone accepts flask, upper cone accepts condenser - Splash head reduces entrainment of raw liquid into the vapour stream
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Distillation Splash Head Adapter
Borosilicate Glass (3.3)
Distillation assemblies in which the condenser is assembled in the vertical mode
19/26 (Condenser), 19/26 (flask)