Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand: Quickfit XA32
47.70 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
- Useful for the connection of dissimilar sized ground glass joints
- Large sized socket at the top and small sized cone at the base
Specifications
19/26 | |
Borosilicate Glass | |
24/29 |
Useful for the connection of dissimilar sized ground Glass joints | |
Expansion adapter |
Product information
File size: 2.8 MB
Product Content Correction
Product Title
Quickfit™ Expansion Socket Adapter > Cone size: 19/26; Socket size: 24/29
Spot an opportunity for improvement?Share a Content Correction